missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging

TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging

Brand:  Molecular Probes™ A24879

Product Code. 15303346

  • 423.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Molecular Probes™ human pluripotent stem cell (PSC) live imaging kits are the only kits offering superior imaging for live human PSCs in one box. This TRA-1-60 Alexa Fluor™ 555 Conjugate Kit includes an optimally labeled primary antibody paired with reduced-background imaging medium (FluoroBrite™ DMEM) for the specific detection of the PSC marker TRA-1-60. For fixed cell staining of PSC markers, we recommend the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit.

With the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging, you can:
  • Confirm the expression of a key pluripotent stem cell marker using an optimally dye-conjugated antibody
  • Preserve cell health and maximize fluorescence signal with FluoroBrite™ DMEM imaging medium
  • Stain live cell samples with confidence using reagents that are tested to be free of common contaminants

    Confirm Expression of a Key PSC Marker
    Powered by the high performance of bright, photostable Alexa Fluor™ dyes, this kit enables reliable live-cell detection of the surface PSC marker TRA-1-60. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM imaging medium.

    Preserve Cell Health and Maximize Signal
    Staining cells in FluoroBrite™ DMEM imaging medium maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.

    Stain Live-cell Samples with Confidence
    Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2μm sterile-filtered and tested to be free of common contaminants.

    What You Get
    Easily scalable to any culture plate, dish, or slide format, the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200 uL staining volume and includes:
  • 200μL of Alexa Fluor™ 555 Mouse Anti-Human TRA-1-60 (50X concentration)
  • 500 mL FluoroBrite™ DMEM
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Alexa Fluor 555
    50 tests
    • 200 μL of Alexa Fluor™ 555 Mouse Anti-Human TRA-1-60 (0.25 mg/mL) (store at 2°C to 8°C)
    • 500 mL FluoroBrite™ DMEM
    • Store at 2°C to 8°C, protect from light
    Conjugate Kit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.