Learn More
Thermo Scientific™ TRACE™ GC Columns for Pesticides
Identify organophosphate and organochlorine pesticides with high performance, sensitivity and reproducibility.
Brand:  Thermo Scientific™ 26RF142F
 View more versions of this product
Product Code. 10211635
  • 932.11 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Identify organophosphate and organochlorine pesticides with high performance, sensitivity, and reproducibility using the Thermo Scientific™ TRACE™ GC Columns for Pesticides. These columns are specifically designed and tested to meet the most stringent requirements for the analysis of pesticides and herbicides.
Excellent Quality and Reproducibility:
  • Low bleed decreases MS contamination
  • Innovative design facilitates use at higher temperatures
  • Column inertness results in minimal peak tailing and decreased breakdown of sensitive samples
Alternative Uses:
  • Chlorinated aromatics
  • Plant sterols
  • Pyrethroids
TRUSTED_SUSTAINABILITY
Specifications
Specifications
280/300°C
0.25 mm
Trace™ Pesticide
Pesticides Analysis
0.25 μm
30 m x 0.25 mm x 0.25 um with 5 m Guard
0.25 mm
TRACE
G43
0.2 μm
30 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.