Learn More
Thermo Scientific™ UHPLC Filters
Protects columns without compromising performance.
220.88 € - 331.19 €
Specifications
Product Line Thermo Scientific
View More Specs
Products 3
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11550944
View Documents
Thermo Scientific™
22185
1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm
331.19 €
Pack of 5
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
10775706
View Documents
Thermo Scientific™
27006
UHPLC Direct Connect Filter Holder
290.79 €
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
10127594
View Documents
Thermo Scientific™
22180
2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm
220.88 €
Pack of 5
In Stock
to see stock.
Add to basket
Description
Description
Sacrificing performance is not an option with Thermo Scientific™ UHPLC filters that protect columns for Hypersil GOLD™ 1.9μm and Syncronis 1.7μm columns. These UHPLC filters provide a low-volume filter cartridge design, maintain peak shape, and ensure minimal efficiency loss through dispersion.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
Thermo Scientific