Learn More
Thermo Scientific™ TRACE™ TR-5 GC Columns
Choose these excellent starting columns for method development, capable of performing most required separations.
134.22 € - 1342.04 €
Specifications
Stationary Phase Trace™ TR-5
USP Type G27, G36
Polarity Non-Polar
View More Specs
Products 17
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
10219844
View Documents
Thermo Scientific™
260E113P
7 m 0.32 mm 0.25 μm N/A N/A N/A
10374603
View Documents
Thermo Scientific™
260E130P
15 m 0.25 mm 0.25 μm
377.88 €
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
10079372
View Documents
Thermo Scientific™
260E131P
15 m 0.32 mm 0.25 μm
395.79 €
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
10243263
View Documents
Thermo Scientific™
260E142P
30 m 0.25 mm 0.25 μm
638.92 €
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
10253263
View Documents
Thermo Scientific™
260E143P
30 m 0.32 mm 0.25 μm
680.75 €
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
10384173
View Documents
Thermo Scientific™
260E154P
60 m 0.25 mm 0.25 μm
1124.47 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10424963
View Documents
Thermo Scientific™
260E155P
60 m 0.32 mm 0.25 μm
1202.05 €
Each
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
10293593
View Documents
Thermo Scientific™
260E223P
30 m 0.25 mm 0.5 μm N/A N/A N/A
10735983
View Documents
Thermo Scientific™
260E224P
30 m 0.32 mm 0.5 μm
680.75 €
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
10532465
View Documents
Thermo Scientific™
260E225P
30 m 0.53 mm 0.5 μm
767.22 €
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
10262005
View Documents
Thermo Scientific™
260E242P
100 m 0.32 mm 0.5 μm
1342.04 €
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
10516765
View Documents
Thermo Scientific™
260E286P
15 m 0.53 mm 1 μm N/A N/A N/A
10096004
View Documents
Thermo Scientific™
260E297P
30 m 0.32 mm 1 μm
700.77 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10089182
View Documents
Thermo Scientific™
260E298P
30 m 0.53 mm 1 μm
767.22 €
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
10107504
View Documents
Thermo Scientific™
260E309P
60 m 0.32 mm 1 μm N/A N/A N/A
10585415
View Documents
Thermo Scientific™
260E336P
30 m 0.53 mm 1.5 μm
747.47 €
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
10223423
View Documents
Thermo Scientific™
260E470P
30 m 0.53 mm 5 μm
745.45 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
Description
Description
Excellent starting columns for method development,Thermo Scientific™ TRACE™ TR-5 GC Columns are capable of performing most required separations.
  • Nonpolar phase, 5% phenyl methyl polysiloxane
  • High operating temperature and extremely low-bleed
  • Widely used in a variety of applications
  • Similar to: DB-5, BP5, Rtx-5, HP-5, Ultra-2, PTE-5, SPB5, MDN-5, CP-Sil 8CB, SPB-5, AT-5, ZB-5, 007-2(MPS-5), SE-52, SE-54
Recommended for:
  • Alcohols
  • Free fatty acids
  • Aromatics
  • Flavors
  • Low polarity pesticides
Specifications
Specifications
Trace™ TR-5
G27, G36
Non-Polar
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ TRACE™ TR-5 GC Columns