missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TRACE™ TR-SimDist GC Columns

Use this nonpolar GC column for the demanding requirements of high temperature simulated distillation analysis.

Brand:  Thermo Scientific™ 260S348P

361.39 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10434803

  • 363.20 € / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For the demanding requirements of high-temperature, simulated distillation analysis, choose the Thermo Scientific™ TRACE™ TR-SimDist Column. The column has a strongly cross-linked phase with an operating temperature limit of 400°C. Its stable low-bleed profile facilitates consistent background subtraction, which is common practice in these analyses.

Engineered for Superior Performance:
  • Nonpolar phase, 100% dimethyl polysiloxane
  • Optimized for simulated distillation analysis
  • High temperature limits (400˚C for films <1.0μm; 370˚C for 2.65μm films)
  • Strongly cross-linked
Applications:
  • High molecular weight hydrocarbons
  • Simulated distillation
  • ASTM D2887, D6532
Recommended For:
  • Simulated distillation
  • Petroleum
Alternative Uses:
  • Xylenes
Similar To:
  • DB-HT Sim Dis
  • DB-2887
  • BPX1
  • Rtx™-2887
  • HP-1
  • Petrocol™ 2887
  • Petrocol™ EX2887
TRUSTED_SUSTAINABILITY
Specifications

Specifications

370°C
Trace™ TR-SimDist
2.65 μm
10m x 0.53mm x 2.65um
0.53 mm
GC/MS
10 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.