Learn More
Fisherbrand™ Traceable™ Memory-Loc™ Data Logging Thermohygrometers with Calibration
The Traceable™ Memory-Loc™ Data Logging Thermohygrometer with Calibration allows you to safely secure collected information — data cannot be overwritten or modified.
Brand:  Fisherbrand™ 98767-16
Product Code. 17367649
  • 381.00 € / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory.
  • Store up to 30 unique alarm events.
  • Smart-Alarm™ with visual/audible alarm continues to alert even if the unit returns to nonalarm conditions.
  • Adjustable sampling rate — from 1 min. to 12 hr. in 1 min. steps.
  • Can upload a USB downloaded file and store on a TraceableLIVE™ cloud account.
Specifications
Specifications
Thermohygrometer
0.1
±0.4°C
688,086 Readings
0 to 95%
-20°C to +70°C
Traceable™ Memory-Loc™ Thermohygrometer with Calibration
14 x 2.5 x 8.3 cm