missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Memory-Loc™ Datalogging Thermometer

Traceable™ Memory-Loc™ Datalogging Thermometer that can be used in 21 CFR 11 environments.

Brand:  Fisherbrand™ 93680-39

 View more versions of this product

Product Code. 15204036

  • 261.00 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Traceable™ Memory-Loc™ Datalogging Thermometer that can be used in 21 CFR 11 environments.

  • Data-logging thermometer with fixed one-minute timing intervals
  • Meets current CDC requirements and additional recommendations for vaccine storage and monitoring
  • Can be used in 21 CFR 11 environments, raw data is locked and cannot be cleared or changed on base unit
  • Stores up to ten unique alarm events
  • Capable of storing over 1 million temperature observations
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, low memory, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Weight: 5 oz.
Hi/Lo Alarms and Time/Date Stamps
  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
USB Data Transferring
  • All new feature allows recorded data (CSV file) to be transferred from thermometer to PC (or Mac™) using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location
  • Raw data on unit cannot be overwritten or modified
  • Indicator light will appear when 95% of memory is full
Data-Logging Features
  • Can be used in environments requiring 21 CFR 11 compliance
  • Thermometer logs two years of observations with a one-minute logging interval
  • Device will stop recording when storage is full and will not overwrite recorded data
  • Data is unable to be deleted from memory once stored
Bottle Probe Features
  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples
  • Meets all current CDC requirements and recommendations for temperature monitoring of vaccine storage as part of the Vaccines for Children program

Order Info

Replacement battery: Cat. No. 1111

Specifications

Specifications

Datalogger
Not Calibrated
2 Probes (Bottle Probe and 5/16 in. dia. x 3/4 in. long)
NIST, ISO/IEC 17025, A2LA, CDC
Temperature-Buffered Bottle Sensor
0.01°
-58°F to +158°F
5 oz.
LCD
LED Indicator, Audible, Flashing LCD Segment
1 Bottle probe with cable and 1 standard Plastic probe of 0.31 in. dia. x 0.75 in. L with cable, A/C adapter, stand, Velcro magnetic strips, wall mount, batteries
USB
±0.25°C
-50°C to +70°C
-72.4°F to +316.4°F
141.7 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers