missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM), Fisherbrand™

Ideal for lab or field applications

31.16 € - 49.70 €

Specifications

Chemical Name or Material Conductivity Standard
Packaging 6/Pk.
Color Clear
For Use With (Equipment) Conductivity Meter
Solubility Soluble in water
View More Specs

Products 6
Product Code Brand Quantity Conductivity Price Quantity & Availability  
Product Code Brand Quantity Conductivity Price Quantity & Availability  
11774246
missing translation for 'viewDocuments'
Fisherbrand
98769-27
6 x 100 mL 100 μ℧
49.70 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
11784246
missing translation for 'viewDocuments'
Fisherbrand
98769-28
100 mL 1000 μ℧
49.70 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
11754246
missing translation for 'viewDocuments'
Fisherbrand
98769-26
6 x 100 mL 1413 μ℧
46.60 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
11794246
missing translation for 'viewDocuments'
Fisherbrand
98769-29
100 mL 10,000 μ℧
49.70 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
11704256
missing translation for 'viewDocuments'
Fisherbrand
98769-30
100 mL 100,000 μ℧
49.70 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
11744246
missing translation for 'viewDocuments'
Fisherbrand
98769-25
6 x 100 mL 10, 100, 1000, 1413, 10,000, 100,000 μ℧ This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Ideal for lab or field applications

Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.

Calibration within container — zero contamination

  • Traceable to NIST (National Institute of Standards and Technology)
  • Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
  • 100% compatible with all conductivity meters
  • A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
  • Includes an individual temperature chart and step-by-step calibration instructions
  • Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
  • Single use Eliminates external contamination concerns Each standard contains 100mL

Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12

Order Info

Each bottle contains 100mL

Specifications

Specifications

Conductivity Standard
Clear
Soluble in water
6/Pk.
Conductivity Meter
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Traceable Conductivity Standard

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.