missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Precision Lollipop™ Water-Resistant Thermometers

This digital and water-resistant thermometer provides results in two seconds

Brand:  Fisherbrand™ 93679-68

 View more versions of this product

Product Code. 16516820

  • 90.85 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Description

Description

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Water-resistant, IP64, case allows for dependable readings even in wet environments
  • Quickly recall min/max readings and switch between °F and °C with the press of a button
  • Read in any position and lighting condition with backlit display that can be rotated up to 180°
  • Choose between an integrated or 9-ft (2.7 m) cabled sensor
Specifications

Specifications

Lollipop Thermometer
Cabled Probe
-58°F to +482°F
20.32 cm
2.54 cm
7.62 cm
0.907 kg
Protective Sheath, Suction-Cup Mount, Hook and Loop Mount, Magnetic Mount, CR2032 Battery, and NIST-Traceable Certificate (with data)
±0.5°C from -20°C to 200°C, ±1°C otherwise
LCD
8 in.
1 in.
3 in.
2 lb.
Temperature Monitoring
NIST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.