missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ TraceableGO™ Bluetooth Datalogging Hygrometer

Fisherbrand TraceableGO™ Bluetooth Datalogging Hygrometer fills in the unknowns between destinations

Brand:  Fisherbrand™ 61010-55

Product Code. 15848946

  • 111.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Batteries, Wall mount, Traceable Certificate

Description

Description

  • Fills in the unknowns between destinations
  • Critical Information (Temperature, Trip Duration, MIN/MAX, Battery Level and Alarms) displayed on LCD at all times
  • Simple, single button use
  • Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location
  • Compatible with TraceableGO or TraceableLIVE Mobile Apps:
    • Once the trip has ended, users may export the data
    • App immediately identifies if the contents experienced conditions outside user-defined parameters
    • View if an alarm was triggered
    • Generate an instant data report
    • Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours
    • Logging duration is 227 days (7.5 months)
  • TraceableGO™ Bluetooth devices are pre-programmed with alarm values

Certifications

ISO 17025, A2LA, NIST

Services

1 year from date of purchase

Specifications

Specifications

Datalogging Hygrometer
0.001
-20°C to +70°C
Hi/Low Temperature and Humidity
LCD
5 sec.
0.1°
2.0 cm
0.17 kg
Batteries, wall mount, Traceable™ Certificate
0 to 95%
±3% RH between 5% and 75%, ±5% RH otherwise
±0.4°C between -10°C to +70°C, otherwise ±0.5°C
2 x AAA
90 Alarm Events, 65,536 Temperature Readings
-4°F to +158°F
8.9 cm
5.1 cm
Temperature and humidity monitoring
ISO 17025, A2LA, NIST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.