missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Critical Information (Temperature, Trip Duration, MIN/MAX, Battery Level and Alarms) displayed on LCD at all times Simple, single button use
- Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location
- Compatible with TraceableGO or TraceableLIVE Mobile Apps:
- Once the trip has ended, users may export the data
- App immediately identifies if the contents experienced conditions outside user-defined parameters
- View if an alarm was triggered
- Generate an instant data report
- Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours
- Logging duration is 227 days (7.5 months)
- TraceableGO™ Bluetooth devices are pre-programmed with alarm values
Specifications
Temperature Datalogger | |
64,000 Readings, 7.5 Months, 5 Minute Interval | |
TraceableLIVE | |
Bluetooth datalogging thermometer with RTD probe | |
1 | |
Bluetooth | |
±0.2°C | |
-130°F to +212°F | |
8.9 x 2 x 5.1 cm | |
5.1 cm | |
8.9 cm | |
2 cm | |
0.17 kg |
Multi-line LCD | |
High/Low | |
2 AAA | |
Batteries, wall mount, Traceable™ Certificate | |
ISO 17025, NIST | |
Temperature monitoring | |
0.1° | |
-90°C to +100°C | |
2 in. | |
3.5 in. | |
0.8 in. | |
6 oz. |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ TraceableGO™ Temperature Data Logger with Bluetooth™, Calibration and Platinum Probe > Traceable Temperature Bluetooth™ Enabled Data Logger Compatible with TraceableGO™ App and TraceableLIVE™ Cloud Service; Probe Type: RTD Probe
Option to subscribe to cloud-based TraceableLIVE™ service to upload data to cloud.
Spot an opportunity for improvement?Share a Content Correction