Learn More
Fisherbrand™ TraceableOne™ 10 Pack Single-User Datalogger
Use logger right out of the box to track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals and food items during transit. Fisherbrand™ TraceableOne™ 10 Pack Single-User Datalogger allows for monitoring materials every step of the way.
169.00 €
Specifications
Type Datalogger
Calibration Calibrated
Software Type No software required, download data as PDF report
Includes Batch-calibrated Traceable certificate
IP Rating IP67
View More Specs
Products 8
Product Code Brand Data Memory Description Alarm Types Price Quantity & Availability  
Product Code Brand Data Memory Description Alarm Types Price Quantity & Availability  
16089511
View Documents
Fisherbrand™
6483
16,000 Readings, 10 Day, 1 Minute Interval Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16059521
View Documents
Fisherbrand™
6493
16,000 Readings, 90 Day, 10 Minute Interval Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16039521
View Documents
Fisherbrand™
6490
16,000 Readings, 60 Day, 6 Minute Interval Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16049521
View Documents
Fisherbrand™
6492
16,000 Readings, 90 Day, 10 Minute Interval Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16009521
View Documents
Fisherbrand™
6486
16,000 Readings, 30 Day, 3 Minute Interval Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16099511
View Documents
Fisherbrand™
6484
16,000 Readings, 10 Day, 1 Minute Interval Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16019521
View Documents
Fisherbrand™
6487
16,000 Readings, 30 Day, 3 Minute Interval Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16029521
View Documents
Fisherbrand™
6489
16,000 Readings, 60 Day, 6 Minute Interval Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Logger generates PDF report
  • Download directly to PC
  • No Software required
  • Compact
  • Single-use
  • Flashing Red and Green LED's indicate if logger is recording data, has stopped recording and if an alarm has been triggered
  • Press start to begin recording crucial temperature data, 30 minute time delay before logging begins
Specifications
Specifications
Datalogger
No software required, download data as PDF report
IP67
±0.5°C from -20° to 40°C, otherwise ±1°C
-30°C to +70°C
Battery-Powered
Calibrated
Batch-calibrated Traceable certificate
Refrigerated transportation samples
0.01°
8.89 cm