Learn More
Thermo Scientific™ TraceGOLD TG-200MS GC Column
Thermo Scientific™ TraceGOLD™ TG-200MS GC Column has an exceptionally inert phase with selectivity and elution order optimized for difficult separations.
Brand:  Thermo Scientific™ 26084-2980
 View more versions of this product
Product Code. 15820900
  • 860.64 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Trifluoropropylmethyl polysiloxane phase resolves compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including ECD, NPD and MS
  • Appropriate for use as a confirmation GC column
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 m
G6
1 μm
0.53 mm
TraceGOLD TG-200MS