Learn More
Thermo Scientific™ TraceGOLD TG-200MS GC Columns
Exceptionally inert mid-polarity columns with selectivity and elution order optimized for difficult separations
Brand: Thermo Scientific™ 26084-2750
This item is not returnable.
View return policy
Description
- Polar phase, trifluoropropylmethyl polysiloxane solid phase resolves compounds that phenyl and cyano phases cannot
- Outstanding thermal stability and low bleed
- Suitable for use with sensitive detectors including ECDs, NPDs and MS
- Confirmation column in combination with another GC column
- Equivalent to USP G6 phase
- Similar to Rtx™-200MS, DB-200, DB-210
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
-20°C to +320/340°C | |
GC Capillary Column | |
Silica Analysis | |
10 m |
32.8 ft. | |
0.15 mm | |
0.15 μm |