Learn More
Thermo Scientific™ TraceGOLD TG-200MS GC Columns
Exceptionally inert mid-polarity columns with selectivity and elution order optimized for difficult separations
Brand:  Thermo Scientific™ 26084-2760
 View more versions of this product
Product Code. 13488018
  • 616.65 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polar phase, trifluoropropylmethyl polysiloxane solid phase resolves compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including ECDs, NPDs and MS
  • Confirmation column in combination with another GC column
  • Equivalent to USP G6 phase
  • Similar to Rtx™-200MS, DB-200, DB-210
TRUSTED_SUSTAINABILITY
Specifications
Specifications
-20°C to +320/340°C
GC Capillary Column
Silica Analysis
20 m
65.61 ft.
0.15 mm
0.15 μm