missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TraceGOLD™ TG-ALC Plus II GC Columns

Perform fast, full separation of blood alcohols and other volatile compounds by headspace gas chromatography using Thermo Scientific™ TraceGOLD™ TG-ALC GC columns.

722.39 € - 744.93 €

Specifications

Product Line TraceGOLD
Max. Temperature 260°C
View More Specs

Products 3
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
15294533
View Documents
Thermo Scientific™
26063-2240
30 m 0.32 mm 0.5 μm
728.36 €
Each
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
 
13456846
View Documents
Thermo Scientific™
26073-2260
30 m 0.53 mm 1.2 μm
722.39 €
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
15234543
View Documents
Thermo Scientific™
26063-2980
30 m 0.53 mm 1 μm
744.93 €
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
Description

Description

Perform fast and full separation of blood alcohols by headspace gas chromatography (GC) using Thermo Scientific™ TraceGOLD™ TG-ALC GC columns. These columns are also ideal for analysis of other volatile compounds including abused inhalant anesthetics, γ-hydroxybutyrate (GHB), γ-butyrolactone (GBL), glycols, and common industrial solvents. TraceGOLD TG-ALC columns have low bleed for excellent signal-to-noise ratio, sensitivity, and mass spectral integrity. Note: Blood alcohol analysis requires dual column confirmation when using GC-FID

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications

Specifications

TraceGOLD
260°C
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.