Learn More
Thermo Scientific™ TraceGOLD™ TG-SQC GC Columns
Ensure the quality of your Thermo Scientific instrument when installed on site.
Brand:  Thermo Scientific™ 26070-1420
 View more versions of this product
Product Code. 10157664
  • 781.35 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure the quality of your Thermo Scientific instrument when installed on site by using the Thermo Scientific™ TraceGOLD™ TG-SQC GC column. The TraceGOLD TG-SQC GC column is optimized for system qualification tests during initial installation, or during service and maintenance of an installed instrument. This column is recommended for benchmark testing only.
Applications:
System qualification tests (Thermo Scientific GC-MS)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 m
Benchmark Testing Only
0.25 μm
0.25 mm
TraceGOLD
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.