Learn More
Thermo Scientific™ TraceGOLD TG-VMS GC Columns
Thermo Scientific™ TraceGOLD TG-VMS GC Columns provide good resolution and fast analysis times for volatile compounds.
Brand:  Thermo Scientific™ 26080-3390
 View more versions of this product
Product Code. 11541154
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ TraceGOLD TG-VMS GC Columns provide good resolution and fast analysis times for volatile compounds.
  • Proprietary stationary phase
  • Optimized dimensions for analysis of volatile organic pollutants
  • Ideally suited for US EPA Method 8260B
  • Similar to Rtx-VMS
Recommended for:
  • Volatile organic pollutants
TRUSTED_SUSTAINABILITY
Specifications
Specifications
260°C
0.32 mm
GC/MS for Volatile Organic Pollutants, GC/MS for US EPA Method 8260B Compounds
30 m
0.32 mm
TraceGOLD™ TG-VMS
1.8 μm
Non-Polar