missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TraceGOLD™ TG-WaxMS A GC Columns

Identify organic acids with high performance, sensitivity and reproducibility while preventing oxidative damage with this low-bleed, polar GC column.

Brand:  Thermo Scientific™ 26087-2850

 View more versions of this product

Product Code. 10754837

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Identify organic acids with high performance, sensitivity and reproducibility while preventing oxidative damage using Thermo ScientificTM TraceGOLDTM TG-WaxMS A GC column.

Engineered for Superior Performance
  • Polar phase, acid-deactivated polyethylene glycol
  • Resists oxidative damage and adsorption of acids
  • Excellent peak shapes for high MW acids
  • Equivalent to USP G25, G35 phases
Applications:
  • Organic acids
  • Free fatty acids
  • Alcohols

A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • This low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.

Recommended For:
  • Acids
  • Carboxylic acids
  • Organic acids
  • Alternative Uses:
  • Alcohols
  • Alcohols in beverages
  • Aldehydes
  • Alditol acetates (sugars)
  • Essential oils
  • Similar To:
  • DB-FFAP
  • HP-FFAP
  • Nukol™
  • OV™-351
  • CP-Wax 58 CB
  • Stabilwax™-DA
  • BP-21
  • Optima™ FFAP
TRUSTED_SUSTAINABILITY
Specifications

Specifications

240/250°C
0.32 mm
G25, G35
1 μm
Polar
0.32 mm
TraceGOLD™ TG-WAXMS A
GC/MS for Organic Acids, GC/MS for Free Fatty Acids, GC/MS for Alcohols
15 m
1umthickness; 0.32mm ID; 15m length
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.