Learn More
trans-1,2-Cyclohexanediol, 98%, Thermo Scientific Chemicals
54.57 € - 167.84 €
Chemical Identifiers
CAS | 1460-57-7 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00063611 |
InChI Key | PFURGBBHAOXLIO-PHDIDXHHSA-N |
Synonym | trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol |
PubChem CID | 92886 |
ChEBI | CHEBI:16931 |
IUPAC Name | (1R,2R)-cyclohexane-1,2-diol |
SMILES | O[C@@H]1CCCC[C@H]1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307136
|
Thermo Scientific Alfa Aesar
L07088.14 |
25 g |
54.57 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317136
|
Thermo Scientific Alfa Aesar
L07088.22 |
100 g |
167.84 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.
Solubility
Soluble in chloroform and methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
1460-57-7 | |
116.16 | |
PFURGBBHAOXLIO-PHDIDXHHSA-N | |
92886 | |
(1R,2R)-cyclohexane-1,2-diol |
C6H12O2 | |
MFCD00063611 | |
trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
CHEBI:16931 | |
O[C@@H]1CCCC[C@H]1O |
Specifications
1460-57-7 | |
231°C to 233°C | |
C6H12O2 | |
3193810 | |
Soluble in chloroform and methanol. | |
O[C@@H]1CCCC[C@H]1O | |
116.16 | |
CHEBI:16931 | |
98% |
101°C to 106°C | |
134°C (273°F) | |
MFCD00063611 | |
trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
PFURGBBHAOXLIO-PHDIDXHHSA-N | |
(1R,2R)-cyclohexane-1,2-diol | |
92886 | |
116.16 | |
trans-1,2-Cyclohexanediol |
Safety and Handling
missing translation for 'einecsNumber' : 215-956-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.