missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Dichloroethylene, 98%, stab. with 4-methoxyphenol, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.
Solubility
Miscible with ethanol, ethyl ether, acetone, benzene and chloroform. Immiscible with water.
Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and bases.
trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.
Solubility
Miscible with ethanol, ethyl ether, acetone, benzene and chloroform. Immiscible with water.
Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and bases.
Specifications
Specifications
Melting Point | -50°C |
Density | 1.252 |
Boiling Point | 47°C to 49°C |
Flash Point | 2°C (35°F) |
Refractive Index | 1.4455 |
Quantity | 100 g |
UN Number | UN1150 |
Beilstein | 1420761 |
Merck Index | 14,92 |
Solubility Information | Miscible with ethanol,ethyl ether,acetone,benzene and chloroform. Immiscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'