Learn More
trans-1,2-Dichloroethylene, 99+%, stabilized, Thermo Scientific Chemicals
81.41 € - 424.11 €
Chemical Identifiers
CAS | 156-60-5 |
---|---|
Molecular Formula | C2H2Cl2 |
Molecular Weight (g/mol) | 96.94 |
MDL Number | MFCD00062942 |
InChI Key | KFUSEUYYWQURPO-OWOJBTEDSA-N |
Synonym | trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform |
PubChem CID | 638186 |
ChEBI | CHEBI:29027 |
IUPAC Name | (E)-1,2-dichloroethene |
SMILES | Cl\C=C\Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10481011
|
Thermo Scientific Acros
406840250 |
25 g | Glass bottle |
81.41 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10558892
|
Thermo Scientific Acros
406842500 |
250 g | Glass bottle |
424.11 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
156-60-5 | |
96.94 | |
KFUSEUYYWQURPO-OWOJBTEDSA-N | |
638186 | |
(E)-1,2-dichloroethene |
C2H2Cl2 | |
MFCD00062942 | |
trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform | |
CHEBI:29027 | |
Cl\C=C\Cl |
Specifications
156-60-5 | |
Colorless | |
48°C | |
Authentic | |
C2H2Cl2 | |
1.4450 to 1.447 | |
01, 188 | |
1.26 | |
Solubility in water: immiscible | |
Cl\C=C\Cl | |
85 to 115ppm MEHQ | |
638186 | |
96.94 | |
Liquid |
-50°C | |
1.2600g/mL | |
6°C | |
99% min. (GC) | |
CHClCHCl | |
MFCD00062942 | |
15, 87 | |
trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform | |
KFUSEUYYWQURPO-OWOJBTEDSA-N | |
(E)-1,2-dichloroethene | |
96.94 | |
CHEBI:29027 | |
99+% | |
trans-1, 2-Dichloroethylene, 99.7% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-860-2
missing translation for 'rtecsNumber' : KV9400000
missing translation for 'tsca' : TSCA
- trans-1,2-Dichloroethylene, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only