missing translation for 'onlineSavingsMsg'
Learn More
trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane, 97%, Thermo Scientific Chemicals Product Code.: 15487466

trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane, 97%, Thermo Scientific Chemicals

Product Code. 15487466
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15487466

Brand: Thermo Scientific Alfa Aesar H62469.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1222709-30-9
Molecular Formula C12H22BrNO2
Molecular Weight (g/mol) 292.217
MDL Number MFCD12827547
InChI Key JPJCPRFAUVDSEA-UHFFFAOYSA-N
Synonym tert-butyl-trans-4-bromomethyl-cyclohexylcarbamate, tert-butyl trans-4-bromomethyl cyclohexyl carbamate, tert-butyl 4-bromomethyl cyclohexylcarbamate, tert-butyl-trans-4-bromomethyl cyclohexylcarbamate, jpjcprfauvdsea-mgcohnpysa-n, tert-butyl n-4-bromomethyl cyclohexyl carbamate, tert-butyl trans-4-bromomethyl cyclohexylcarbamate, trans-1-boc-amino-4-bromomethyl cyclohexane, tert-butyl n-1r,4r-4-bromomethyl cyclohexyl carbamateShow MoreShow Less
PubChem CID 67470265
IUPAC Name tert-butyl N-[4-(bromomethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CBr

Specifications

Quantity 250 mg
Formula Weight 292.21
Percent Purity 97%
Chemical Name or Material trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS01J015 Certificate TypeCertificate of Analysis Date08/06/2023 Catalog NumberH62469.MD
Lot NumberP03I031 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH62469.MD,H62469.03
Lot NumberM09F054 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62469.MD,H62469.03
Lot NumberM20H024 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62469.MD,H62469.03
Lot NumberD02Z012 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62469.MD,H62469.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.