missing translation for 'onlineSavingsMsg'
Learn More
trans-1-(Boc-amino)-4-cyanocyclohexane, 97%, Thermo Scientific Chemicals Product Code.: 15418648

trans-1-(Boc-amino)-4-cyanocyclohexane, 97%, Thermo Scientific Chemicals

Product Code. 15418648
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15418648

Brand: Thermo Scientific Alfa Aesar H62060.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873537-32-7
Molecular Formula C12H20N2O2
Molecular Weight (g/mol) 224.304
MDL Number MFCD11707221
InChI Key QHZJRHRMFIDRFL-UHFFFAOYSA-N
Synonym tert-butyl 4-cyanocyclohexyl carbamate, tert-butyl cis-4-cyanocyclohexylcarbamate, 4-cyano-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-cyanocyclohexylcarbamate, tert-butyl n-4-cyanocyclohexyl carbamate, tert-butyl 1r,4r-4-cyanocyclohexyl carbamate, qhzjrhrmfidrfl-mgcohnpysa-n, trans-1-boc-amino-4-cyanocyclohexane, trans tert-butyl 4-cyanocyclohexyl carbamate
PubChem CID 46172880
IUPAC Name tert-butyl N-(4-cyanocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C#N

Specifications

UN Number UN3439
Quantity 1 g
Formula Weight 224.29
Percent Purity 97%
Chemical Name or Material trans-1-(Boc-amino)-4-cyanocyclohexane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC29Z063 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog NumberH62060.03
Lot NumberC29Z063 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62060.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-1-(Boc-amino)-4-cyanocyclohexane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.