Learn More
trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
58.99 € - 791.00 €
Chemical Identifiers
CAS | 1013-96-3 |
---|---|
Molecular Formula | C9H7NO4 |
Molecular Weight (g/mol) | 193.158 |
MDL Number | MFCD00007189 |
InChI Key | BBQDLDVSEDAYAA-AATRIKPKSA-N |
Synonym | 2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668 |
PubChem CID | 735923 |
SMILES | C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11434564
|
Thermo Scientific Alfa Aesar
A13171.14 |
25 g |
58.99 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444564
|
Thermo Scientific Alfa Aesar
A13171.22 |
100 g |
188.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454564
|
Thermo Scientific Alfa Aesar
A13171.36 |
500 g |
791.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1013-96-3 | |
193.158 | |
BBQDLDVSEDAYAA-AATRIKPKSA-N | |
735923 |
C9H7NO4 | |
MFCD00007189 | |
2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668 | |
C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-] |
Specifications
1013-96-3 | |
Odorless | |
MFCD00007189 | |
2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668 | |
C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-] | |
735923 | |
98% |
245°C to 249°C | |
C9H7NO4 | |
2211209 | |
BBQDLDVSEDAYAA-AATRIKPKSA-N | |
193.158 | |
193.16 | |
trans-2-Nitrocinnamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-309-4
missing translation for 'rtecsNumber' : UD3642850
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only