missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octenal, 95%, Thermo Scientific Chemicals

Product Code. 11413108
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in flavoring tangerine, mandarin, cherry, melon and orange and also as a fragrances agent. It causes defects for locomotion in green fluorescent protein (GFP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in flavoring tangerine, mandarin, cherry, melon and orange and also as a fragrances agent. It causes defects for locomotion in green fluorescent protein (GFP). It is an antimicrobial agent that destroys fungi by suppressing their ability to grow or reproduce.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils.

Notes
Air Sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2548-87-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00007011
InChI Key LVBXEMGDVWVTGY-VOTSOKGWSA-N
Synonym trans-2-octenal, e-oct-2-enal, e-2-octenal, 2-octenal, trans-2-octen-1-al, e-2-octen-1-al, 2-octen-1-al, 2-octenel, 2-octenal, e, 2-octenal, 2e
PubChem CID 5283324
ChEBI CHEBI:61748
IUPAC Name (E)-oct-2-enal
SMILES CCCCCC=CC=O

Specifications

Density 0.84
Boiling Point 84°C to 86°C (19 mmHg)
Flash Point 71°C (160°F)
Odor Pleasant
Refractive Index 1.45
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils.
Formula Weight 126.2
Percent Purity 95%
Chemical Name or Material trans-2-Octenal
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'