missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Undecenal, 95%, remainder mainly cis-isomer, Thermo Scientific™

Product Code. 11392516
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53448-07-0
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00014680
InChI Key PANBRUWVURLWGY-MDZDMXLPSA-N
Synonym trans-2-undecenal, 2-undecenal, e-undec-2-enal, e-2-undecenal, 2-undecenal, 2e, 2-undecen-1-al, 2e-2-undecenal, trans-2-undecen-1-al, 2-undecenal, e, unii-fnp3s9mg30
PubChem CID 5283356
IUPAC Name (E)-undec-2-enal
SMILES CCCCCCCCC=CC=O

Specifications

Density 0.844
Boiling Point 115°C (10 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.457
Quantity 25 g
Beilstein 1762239
Sensitivity Air sensitive
Formula Weight 168.28
Percent Purity 95%
Assay remainder mainly cis-isomer
Chemical Name or Material trans-2-Undecenal
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'