missing translation for 'onlineSavingsMsg'
Learn More
trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl, 99%, Thermo Scientific™ Product Code.: 15432896

trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl, 99%, Thermo Scientific™

Product Code. 15432896
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15432896

Brand: Thermo Scientific Alfa Aesar H56790.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132123-39-8
Molecular Formula C21H23F3
Molecular Weight (g/mol) 332.41
MDL Number MFCD09839001
InChI Key RRKRBRVTKIRLHH-UHFFFAOYSA-N
Synonym 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 4'-trans-4-propylcyclohexyl-3,4,5-trifluorobiphenyl, 1,2,3-trifluoro-5-4-4-propylcyclohexyl phenyl benzene, 4'-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, 3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, trans-3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, 4-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, trans-3,4,5-trifluoro-4'-4-n-propylcyclohexyl biphenyl, 1,1'-biphenyl, 3,4,5-trifluoro-4'-trans-4-propylcyclohexylShow MoreShow Less
PubChem CID 15014921
IUPAC Name 1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)phenyl]benzene
SMILES CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C(=C3)F)F)F

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 332.4
Percent Purity 99%
Chemical Name or Material trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBF120551 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.