missing translation for 'onlineSavingsMsg'
Learn More
trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl, 97%, Thermo Scientific™ Product Code.: 15452066

trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl, 97%, Thermo Scientific™

Product Code. 15452066
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15452066

Brand: Thermo Scientific Alfa Aesar H55359.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Sparingly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85312-59-0
Molecular Formula C21H24F2
Molecular Weight (g/mol) 314.42
MDL Number MFCD09838999
InChI Key VULXHDGYVHCLLN-UHFFFAOYSA-N
Synonym 4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl
PubChem CID 612306
IUPAC Name 1,2-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
SMILES CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1

Specifications

Melting Point 66°C to 68°C
Quantity 1 g
Solubility Information Sparingly soluble in water.
Formula Weight 314.42
Percent Purity 97%
Chemical Name or Material trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.