missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Fluorocinnamic acid, 98%, Thermo Scientific Chemicals

Product Code. 11475746
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20595-30-6
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.151
MDL Number MFCD00004383
InChI Key RTSIUKMGSDOSTI-SNAWJCMRSA-N
Synonym 3-fluorocinnamic acid, 3-3-fluorophenyl acrylic acid, trans-3-fluorocinnamic acid, 2e-3-3-fluorophenyl prop-2-enoic acid, e-3-3-fluorophenyl acrylic acid, m-fluorocinnamic acid, trans 3-fluorocinnamic acid, 3-3-fluoro-phenyl-acrylic acid, trans-m-fluorocinnamic, e-3-3-fluorophenyl prop-2-enoic acid
PubChem CID 1551219
IUPAC Name (E)-3-(3-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)F)C=CC(=O)O

Specifications

Melting Point 165°C to 168°C
Odor Odorless
Quantity 25 g
Beilstein 1365172
Formula Weight 166.15
Percent Purity 98%
Chemical Name or Material trans-3-Fluorocinnamic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'