missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(4-n-Propylcyclohexyl)cyclohexanone, 97%, Thermo Scientific™

Product Code. 15481608
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as pharmaceutical intermediate.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82832-73-3
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.372
MDL Number MFCD04038797
InChI Key AKCZQKBKWXBJOF-UHFFFAOYSA-N
Synonym 4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509
PubChem CID 4354185
IUPAC Name 4-(4-propylcyclohexyl)cyclohexan-1-one
SMILES CCCC1CCC(CC1)C2CCC(=O)CC2

Specifications

Quantity 1 g
Formula Weight 222.37
Percent Purity 97%
Chemical Name or Material trans-4-(4-n-Propylcyclohexyl)cyclohexanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'