missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanol, 97%, Thermo Scientific Chemicals

Product Code. 10133441
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27489-62-9
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
InChI Key IMLXLGZJLAOKJN-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanol, 4-aminocyclohexanol, cis-4-aminocyclohexanol, cis-4-amino-cyclohexanol, cyclohexanol, 4-amino-, trans, 1r,4r-4-aminocyclohexan-1-ol, cyclohexanol, 4-amino, trans-4-hydroxycyclohexylamine, 1r,4r-4-aminocyclohexanol, aminocyclohexanol trans-4
PubChem CID 81293
IUPAC Name 4-aminocyclohexan-1-ol
SMILES C1CC(CCC1N)O

Specifications

Melting Point 108.0°C to 113.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 115.17
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material trans-4-Aminocyclohexanol
compliance-icons
Product Identifier
  • trans-4-Aminocyclohexanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'