Learn More
trans-4-Aminocyclohexanol hydrochloride, 97%, Thermo Scientific Chemicals
28.50 € - 253.00 €
Chemical Identifiers
CAS | 50910-54-8 |
---|---|
Molecular Formula | C6H14ClNO |
Molecular Weight (g/mol) | 151.63 |
MDL Number | MFCD00012566,MFCD07366531,MFCD06410647 |
InChI Key | RKTQEVMZBCBOSB-UHFFFAOYSA-N |
Synonym | trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl |
PubChem CID | 522619 |
IUPAC Name | 4-aminocyclohexan-1-ol;hydrochloride |
SMILES | Cl.NC1CCC(O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11426057
|
Thermo Scientific Alfa Aesar
B25070.06 |
5 g |
28.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11436057
|
Thermo Scientific Alfa Aesar
B25070.14 |
25 g |
85.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11446057
|
Thermo Scientific Alfa Aesar
B25070.22 |
100 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
50910-54-8 | |
151.63 | |
RKTQEVMZBCBOSB-UHFFFAOYSA-N | |
522619 | |
Cl.NC1CCC(O)CC1 |
C6H14ClNO | |
MFCD00012566,MFCD07366531,MFCD06410647 | |
trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl | |
4-aminocyclohexan-1-ol;hydrochloride |
Specifications
50910-54-8 | |
C6H14ClNO | |
3909294 | |
trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl | |
Cl.NC1CCC(O)CC1 | |
151.63 | |
151.64 | |
trans-4-Aminocyclohexanol hydrochloride |
225°C to 230°C | |
MFCD00012566,MFCD07366531,MFCD06410647 | |
Hygroscopic | |
RKTQEVMZBCBOSB-UHFFFAOYSA-N | |
4-aminocyclohexan-1-ol;hydrochloride | |
522619 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.