missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 15428868
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

trans-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181308-57-6
Molecular Formula C12H21NO3
Molecular Weight (g/mol) 227.304
MDL Number MFCD03844598
InChI Key GPDBIGSFXXKWQR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-formylcyclohexylcarbamate, tert-butyl cis-4-formylcyclohexylcarbamate, trans-boc-4-aminocyclohexanecarbaldehyde, tert-butyl n-4-formylcyclohexyl carbamate, trans-4-formyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-formylcyclohexyl carbamate, tert-butyl 4-formylcyclohexyl carbamate, trans-tert-butyl 4-formylcyclohexyl carbamate, tert-butyl 1r,4r-4-formylcyclohexylcarbamate, cis-4-boc-amino cyclohexanecarboxaldehyde
PubChem CID 11020615
IUPAC Name tert-butyl N-(4-formylcyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C=O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 227.3
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-amino)cyclohexanecarboxaldehyde
compliance-icons
Product Identifier
  • trans-4-(Boc-amino)cyclohexanecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : trans-4-(Boc-amino)cyclohexanecarboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.