missing translation for 'onlineSavingsMsg'
Learn More
trans-4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals Product Code.: 11491588

trans-4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals

Product Code. 11491588
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11491588

Brand: Thermo Scientific Alfa Aesar A13890.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.

Solubility
Soluble in alcohol and hot water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-98-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004400
InChI Key KSEBMYQBYZTDHS-HWKANZROSA-N
Synonym ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
PubChem CID 445858
ChEBI CHEBI:17620
IUPAC Name (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(\C=C\C(O)=O)=CC=C1O

Specifications

Melting Point 170°C to 175°C
Quantity 25 g
Beilstein 1371483
Merck Index 14,4062
Solubility Information Soluble in alcohol and hot water.
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material trans-4-Hydroxy-3-methoxycinnamic acid
compliance-icons
Product Identifier
  • trans-4-Hydroxy-3-methoxycinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : (E)-4-Hydroxy-3-methoxycinnamic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.