missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals trans-4-Hydroxy-L-proline, 99+%

64.42 € - 1123.59 €

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PubChem CID 5810
ChEBI CHEBI:18095
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10326470
SDS View Documents
Thermo Scientific Chemicals
121780100
10 g Glass bottle
64.42 €
10g
Please to purchase this item. Need a web account? Register with us today!
10607662
SDS View Documents
Thermo Scientific Chemicals
121780250
25 g Glass bottle
100.50 €
25g
Please to purchase this item. Need a web account? Register with us today!
10336470
SDS View Documents
Thermo Scientific Chemicals
121781000
100 g Glass bottle
285.42 €
100g
Please to purchase this item. Need a web account? Register with us today!
11926601
SDS View Documents
Thermo Scientific Chemicals
121785000
500 g Glass bottle
1123.59 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51-35-4
131.13
trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
CHEBI:18095
C1C(CNC1C(=O)O)O
C5H9NO3
PMMYEEVYMWASQN-DMTCNVIQSA-N
5810
(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid

Specifications

51-35-4
White
Authentic
C5H9NO3
15,4878
− 75.50
Solubility in water: 357.8g/L (20° c). Other solubilities: slightly soluble in alcohol, insoluble in ether
C1C(CNC1C(=O)O)O
95% min. (c=10, H2O, 430nm) 1cm cell
5810
131.13
Crystalline Powder or Crystals
274.0°C to 275.0°C
0.2% (105°C, 3 hrs) max.
99+%
22,191
- 75.50 (20.00°C c=5,H2O)
trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PMMYEEVYMWASQN-DMTCNVIQSA-N
(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
131.13
CHEBI:18095
99+%
trans-4-Hydroxy-L-proline
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-091-9

Product Identifier
  • trans-4-Hydroxy-L-proline

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.