Learn More
trans-4-Hydroxy-L-proline, 99+%, Thermo Scientific Chemicals
Beschreibung
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.

Spezifikation
Spezifikation
Melting Point | 270°C to 273°C (decomposition) |
Beilstein | 471933 |
Merck Index | 14,4840 |
Solubility Information | Soluble in water. |
Quantity | 10 g |
Formula Weight | 131.13 |
Optical Rotation | −76° (c=4 in water) |
Percent Purity | ≥99% |
Chemical Name or Material | trans-4-Hydroxy-L-proline |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y08K019 | Certificate of Analysis | 14/11/2024 | A11851.14,A11851.22,A11851.09 |
Y18I020 | Certificate of Analysis | 24/10/2023 | A11851.14,A11851.22,A11851.09 |
10228248 | Certificate of Analysis | 10/07/2023 | A11851.14,A11851,A11851.22,A11851.09 |
10244334 | Certificate of Analysis | 01/11/2022 | A11851.14,A11851.22,A11851.09 |
P14I015 | Certificate of Analysis | 12/04/2022 | A11851.14,A11851.22,A11851.09 |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.