missing translation for 'onlineSavingsMsg'
Learn More
trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals Product Code.: 11481548

trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals

Product Code. 11481548
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11481548

Brand: Thermo Scientific Alfa Aesar A13870.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.

Solubility
Soluble in ethanol, ethyl acetate and other organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 943-89-5
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00004398
InChI Key AFDXODALSZRGIH-QPJJXVBHSA-N
Synonym 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acidShow MoreShow Less
PubChem CID 699414
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=C(\C=C\C(O)=O)C=C1

Specifications

Melting Point 171°C to 175°C
Quantity 250 g
Beilstein 510468
Solubility Information Soluble in ethanol,ethyl acetate and other organic solvents.
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material trans-4-Methoxycinnamic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10137632 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13870.18,A13870.30,A13870.09
Lot Number10179420 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13870.18,A13870.30,A13870.09
Lot Number10034166 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13870.18,A13870.30,A13870.09
Lot Number10194202 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13870.18,A13870.30,A13870.09
Lot Number10234949 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13870.18,A13870.30,A13870.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.