Learn More
trans-Crotonyl chloride, 90%, tech., Thermo Scientific Chemicals
29.65 € - 933.00 €
Chemical Identifiers
CAS | 625-35-4 |
---|---|
Molecular Formula | C4H5ClO |
Molecular Weight (g/mol) | 104.54 |
MDL Number | MFCD00000734 |
InChI Key | RJUIDDKTATZJFE-NSCUHMNNSA-N |
Synonym | crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride |
PubChem CID | 643131 |
IUPAC Name | (E)-but-2-enoyl chloride |
SMILES | CC=CC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10246010
|
Thermo Scientific Acros
155910050 |
5 g | Glass bottle |
29.65 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10256010
|
Thermo Scientific Acros
155911000 |
100 g | Glass bottle |
98.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321344
|
Thermo Scientific Acros
155915000 |
500 g | Glass bottle |
252.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10311344
|
Thermo Scientific Acros
155910025 |
2.5 kg | Glass Bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
625-35-4 | |
104.54 | |
RJUIDDKTATZJFE-NSCUHMNNSA-N | |
643131 | |
CC=CC(=O)Cl |
C4H5ClO | |
MFCD00000734 | |
crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride | |
(E)-but-2-enoyl chloride |
Specifications
625-35-4 | |
1.0900g/mL | |
35°C | |
90% | |
CH3CH=CHCOCl | |
MFCD00000734 | |
13,208 | |
crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride | |
RJUIDDKTATZJFE-NSCUHMNNSA-N | |
(E)-but-2-enoyl chloride | |
643131 | |
90% | |
Liquid |
Colorless to Brown | |
120°C to 123°C | |
Authentic | |
C4H5ClO | |
1.4585 to 1.4615 | |
02, 411 | |
1.09 | |
Solubility in water: decomposes | |
CC=CC(=O)Cl | |
104.54 | |
104.54 | |
Technical | |
trans-Crotonyl chloride, tech., 90% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-889-9
missing translation for 'tsca' : TSCA
- trans-Crotonyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only