Learn More
trans-Diamminedichloropalladium(II), Premion™, 99.95% (metals basis), Pd 49.9% min, Thermo Scientific Chemicals
POA
Chemical Identifiers
CAS | 14323-43-4 |
---|---|
Molecular Formula | Cl2H6N2Pd |
Molecular Weight (g/mol) | 211.38 |
MDL Number | MFCD00011621 |
InChI Key | WIEZTXFTOIBIOC-UHFFFAOYSA-L |
Synonym | diammine dichloropalladium, trans-diamminedichloropalladium ii, transpalladin, cl2pd.2nh3, trans-bisamminedichloropalladium, palladium chloride diamine, trans-dichlorodiammine palladium ii, trans-diamminedichloropalladium ii , premion |
PubChem CID | 61732 |
IUPAC Name | azane;dichloropalladium |
SMILES | N.N.[Cl-].[Cl-].[Pd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351668
|
Thermo Scientific Alfa Aesar
011037.02 |
0.5 g |
POA
0.50g |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11361668
|
Thermo Scientific Alfa Aesar
011037.09 |
10 g |
POA
10g |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14323-43-4 | |
211.38 | |
WIEZTXFTOIBIOC-UHFFFAOYSA-L | |
61732 | |
N.N.[Cl-].[Cl-].[Pd++] |
Cl2H6N2Pd | |
MFCD00011621 | |
diammine dichloropalladium, trans-diamminedichloropalladium ii, transpalladin, cl2pd.2nh3, trans-bisamminedichloropalladium, palladium chloride diamine, trans-dichlorodiammine palladium ii, trans-diamminedichloropalladium ii , premion | |
azane;dichloropalladium |
Specifications
14323-43-4 | |
Crystalline Powder | |
MFCD00011621 | |
diammine dichloropalladium, trans-diamminedichloropalladium ii, transpalladin, cl2pd.2nh3, trans-bisamminedichloropalladium, palladium chloride diamine, trans-dichlorodiammine palladium ii, trans-diamminedichloropalladium ii , premion | |
WIEZTXFTOIBIOC-UHFFFAOYSA-L | |
azane;dichloropalladium | |
61732 | |
99.95% | |
Pd 49.9% min. | |
(metals basis) |
Yellow | |
Cl2H6N2Pd | |
UN3077 | |
Slightly soluble in water; Soluble in NH4OH; Insoluble in chloroform, acetone | |
N.N.[Cl-].[Cl-].[Pd++] | |
211.38 | |
211.37 | |
Premion™ | |
2.5g/mL | |
trans-Diamminedichloropalladium(II) |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 238-269-3
missing translation for 'rtecsNumber' : RT3560000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only