missing translation for 'onlineSavingsMsg'
Learn More

trans-Styrylacetic acid, 96%, Thermo Scientific Chemicals

41.71 € - 184.92 €

Chemical Identifiers

CAS 1914-58-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00002783
InChI Key PSCXFXNEYIHJST-QPJJXVBHSA-N
Synonym trans-styrylacetic acid, e-4-phenylbut-3-enoic acid, 3e-4-phenylbut-3-enoic acid, 4-phenyl-3-butenoic acid, 3-butenoic acid, 4-phenyl, 3-butenoic acid, 4-phenyl-, 3e, styrylacetic acid, e-4-phenyl-3-butenoic acid, e-styrylacetic acid
PubChem CID 5370625
SMILES OC(=O)C\C=C\C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10414155
SDS View Documents
Thermo Scientific Acros
168100010
1 g Glass bottle
41.71 €
1g
Please to purchase this item. Need a web account? Register with us today!
10205600
SDS View Documents
Thermo Scientific Acros
168100100
10 g Glass bottle
184.92 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1914-58-5
162.19
PSCXFXNEYIHJST-QPJJXVBHSA-N
5370625
C10H10O2
MFCD00002783
trans-styrylacetic acid, e-4-phenylbut-3-enoic acid, 3e-4-phenylbut-3-enoic acid, 4-phenyl-3-butenoic acid, 3-butenoic acid, 4-phenyl, 3-butenoic acid, 4-phenyl-, 3e, styrylacetic acid, e-4-phenyl-3-butenoic acid, e-styrylacetic acid
OC(=O)C\C=C\C1=CC=CC=C1

Specifications

1914-58-5
Authentic
C10H10O2
MFCD00002783
trans-styrylacetic acid, e-4-phenylbut-3-enoic acid, 3e-4-phenylbut-3-enoic acid, 4-phenyl-3-butenoic acid, 3-butenoic acid, 4-phenyl, 3-butenoic acid, 4-phenyl-, 3e, styrylacetic acid, e-4-phenyl-3-butenoic acid, e-styrylacetic acid
OC(=O)C\C=C\C1=CC=CC=C1
162.19
162.19
Crystalline Powder or Needles
82°C to 88°C
95% min. (HPLC)
C6H5CH=CHCH2CO2H
09, 612
PSCXFXNEYIHJST-QPJJXVBHSA-N
(3E)-4-phenylbut-3-enoic acid
5370625
96%
trans-Styrylacetic acid, 96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • trans-Styrylacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.