Learn More
trans,trans-1,3-Butadiene-1,4-dicarboxylic acid, 98%, Thermo Scientific Chemicals
29.20 € - 83.60 €
Chemical Identifiers
CAS | 3588-17-8 |
---|---|
Molecular Formula | C6H6O4 |
Molecular Weight (g/mol) | 142.11 |
MDL Number | MFCD00002702 |
InChI Key | TXXHDPDFNKHHGW-ZPUQHVIOSA-N |
Synonym | trans,trans-muconic acid, muconic acid, e,e-muconic acid, 2,4-hexadienedioic acid, trans, trans-muconic acid, hexa-2,4-dienedioic acid, 2e,4e-hexa-2,4-dienedioic acid, 1,3-butadiene-1,4-dicarboxylic acid, 2,4-hexadienedioic acid, e,e, trans,trans-2,4-hexadienedioic acid |
PubChem CID | 5356793 |
ChEBI | CHEBI:27036 |
IUPAC Name | (2E,4E)-hexa-2,4-dienedioic acid |
SMILES | C(=CC(=O)O)C=CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386736
|
Thermo Scientific Alfa Aesar
L03987.03 |
1 g |
29.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396736
|
Thermo Scientific Alfa Aesar
L03987.06 |
5 g |
83.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
trans,trans-1,3-Butadiene-1,4-dicarboxylic acid is a metabolite found in urine that determines biological exposure index for workers exposed to benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans,trans-1,3-Butadiene-1,4-dicarboxylic acid is a metabolite found in urine that determines biological exposure index for workers exposed to benzene.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
3588-17-8 | |
142.11 | |
TXXHDPDFNKHHGW-ZPUQHVIOSA-N | |
5356793 | |
(2E,4E)-hexa-2,4-dienedioic acid |
C6H6O4 | |
MFCD00002702 | |
trans,trans-muconic acid, muconic acid, e,e-muconic acid, 2,4-hexadienedioic acid, trans, trans-muconic acid, hexa-2,4-dienedioic acid, 2e,4e-hexa-2,4-dienedioic acid, 1,3-butadiene-1,4-dicarboxylic acid, 2,4-hexadienedioic acid, e,e, trans,trans-2,4-hexadienedioic acid | |
CHEBI:27036 | |
C(=CC(=O)O)C=CC(=O)O |
Specifications
3588-17-8 | |
C6H6O4 | |
1722248 | |
trans,trans-muconic acid, muconic acid, e,e-muconic acid, 2,4-hexadienedioic acid, trans, trans-muconic acid, hexa-2,4-dienedioic acid, 2e,4e-hexa-2,4-dienedioic acid, 1,3-butadiene-1,4-dicarboxylic acid, 2,4-hexadienedioic acid, e,e, trans,trans-2,4-hexadienedioic acid | |
TXXHDPDFNKHHGW-ZPUQHVIOSA-N | |
(2E,4E)-hexa-2,4-dienedioic acid | |
5356793 | |
142.11 | |
trans,trans-1,3-Butadiene-1,4-dicarboxylic acid |
∽300°C (decomposition) | |
MFCD00002702 | |
14,6299 | |
Soluble in dimethyl sulfoxide and methanol. | |
C(=CC(=O)O)C=CC(=O)O | |
142.11 | |
CHEBI:27036 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 222-724-8
missing translation for 'rtecsNumber' : MM2235070
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only