missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin (synthetic), 97+%, Thermo Scientific Chemicals

Product Code. 10480711
Click to view available options
Quantity:
10 mg
100 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637-39-4
Molecular Formula C10H13N5O
Molecular Weight (g/mol) 219.25
MDL Number MFCD00213654
InChI Key UZKQTCBAMSWPJD-FARCUNLSSA-N
Synonym trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e
PubChem CID 449093
ChEBI CHEBI:16522
IUPAC Name (E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
SMILES CC(=CCNC1=NC=NC2=C1NC=N2)CO

Specifications

Melting Point 207°C
Color Colorless to Yellow
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 10315
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and dimethylformamide
Spectrophotometry 96.0% min. (on dried substance)
Formula Weight 219.25
Percent Purity 97+%
Related Substances 2% max. (HPLC)
Physical Form Liquid
Chemical Name or Material trans-Zeatin (synthetic), 97%
Show More Show Less
Product Identifier
  • trans-Zeatin (synthetic)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'