missing translation for 'onlineSavingsMsg'
Learn More

Merck Transducer Protector

Transducer Protector for applications requiring super hydrophobic membranes.

Brand:  Merck SLGVS25PS

 View more versions of this product

Product Code. 10410221

  • 237.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Hydrophobic Polyvinylidene Fluoride (PVDF)
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Transducer Protector
PVDF
25 mm
Clear Inlet, Blue Outlet
50 Pk.
Dualex Plus
2.5 mm
100 mL
Inlet/Outlet: Female Luer-Lok/Male Luer slip
A 25 mm diameter sterile syringe filter with a 0.22 μm pore size Hydrophobic PVDF membrane. Comes in a pack of 50.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.