missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Transferpette™ -12 Electronic Microliter Pipette

Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.

Brand:  Brand™ 705453

717.34 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10131932

  • 1280.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Functional, ergonomic housing design
  • 12 channels
  • Individually adjustable finger rest
  • Intuitive menu structure and comprehensively illustrated user manual
  • Significantly reduced tip attachment and ejection forces using universal tip
  • Corrosion-resistant piston and ejector
  • Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis, dispensing
  • Ready for use: 4000 pipetting cycles with each battery charge, battery refresh function even during recharging
  • Each pipette has a DE-M marking and is supplied with performance certificate
  • Supplied with battery, AC adapter, device stand, reagent reservoir, mounting tool, silicone oil and one set of sealing rings made of FKM

TRUSTED_SUSTAINABILITY
Specifications

Specifications

≤ 0.8%, ± 0.8μL
Autoclavable
12
Yes
5 to 100 μL
Ergonomic
0.1μL
230V/50 Hz
50 Hz
360°
Yellow
Transferpette™
Electronic Pipette
LCD
Europe AC Adapter
1 Each
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers