missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Transferpette™ Electronic Pipette

Combines the globally proven properties of mechanical piston operated air displacement pipettes with the advantages of an electronic device.

Brand:  BRAND™ 705303

307.35 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10704733

  • 549.00 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The pipette shaft can be unscrewed and is entirely autoclavable (121°C)
  • Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis and dispensing
  • Easy Calibration-Technique for making quick adjustments without tools
  • CE-marked according to IVD-Directive 98/79 EG
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±1.6μL
1
Electronic Pipette
EU
Autoclavable
Transferpette™
10 to 200 μL
15°C to 40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers