missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Transferpette™ Electronic Pipette

Combines the globally proven properties of mechanical piston operated air displacement pipettes with the advantages of an electronic device.

Brand:  BRAND™ 705309

307.35 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 12308593

  • 549.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The pipette shaft can be unscrewed and is entirely autoclavable (121°C)
  • Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis and dispensing
  • Easy Calibration-Technique for making quick adjustments without tools
  • CE-marked according to IVD-Directive 98/79 EG
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.1μL
1
Electronic Pipette
UK/Ireland
Autoclavable
Transferpette™
0.5 to 10 μL
15°C to 40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.