missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Translucent LDPE Bottles

Product Code. 11513242 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Pack of 6
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 11513242

Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Use these bottles for a variety of general storage applications. Fisherbrand™ Translucent HDPE Bottles are leak-proof and offer good chemical resistance.

  • Clear general purpose bottles
  • Blue polypropylene screw closure

Specifications

Capacity (Metric) 1000 mL
Color Translucent
Includes Blue Polypropylene screw closure
Material LDPE
No. per Pack 12 Pack
Closure Color Blue
Closure Included Yes
Mouth Wide
Height (Metric) 140 mm
Product Type Wide Mouth Bottle
Closure Material Polypropylene
Closure Size 70/R3
Diameter (Metric) 90 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.