missing translation for 'onlineSavingsMsg'
Learn More
Fisherbrand™ Translucent Bottles, MDPE Product Code.: 11523242
In Stock
to see stock.
Add to basket

Fisherbrand™ Translucent Bottles, MDPE

Product Code. 11523242 Shop All Fisherbrand Products
30 mL
Click to view available options
Capacity (Metric):
30 mL
125 mL
150 mL
250 mL
500 mL
1000 mL

Product Code. 11523242

In Stock
to see stock.
Add to basket
Add to basket

Use these general purpose bottles for liquid storage and sampling. Fisherbrand™ Translucent Bottles are leakproof and offer excellent chemical resistance. Autoclavable with cap loosened Ideal for general laboratory applications.

  • Clear general purpose bottles
  • Polypropylene screw closure

Specifications

Capacity (Metric) 30 mL
Color Translucent
For Use With (Application) Ideal for general laboratory applications
Includes Polypropylene screw closure
Material MDPE
No. per Pack 5
Closure Included Yes
Autoclavable Autoclavable
Mouth Extra Wide
Height (Metric) 86 mm
Product Type General Purpose Bottle
Closure Material Polypropylene
Closure Size 33/R3
Diameter (Metric) 36 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Translucent Bottles, MDPE >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.