missing translation for 'onlineSavingsMsg'
Learn More

Transparent Glass Staining Trough

Prevent evaporation of staining liquid using the drop-on lid supplied with the Transparent Glass Staining Trough. This heavy glass staining jar is an essential microscope accessory for the laboratory.

16.05 € - 93.15 €

Specifications

Closure Included Closure Included
Bottom Shape Flat
View More Specs

Products 3
Product Code Brand Height (Metric) Type Price Quantity & Availability  
Product Code Brand Height (Metric) Type Price Quantity & Availability  
12858735
View Documents
N/A
2959
110 mm Coplin Staining Jar
50.10 €
Each
In Stock
to see stock.
Edge
Add to basket
 
12868735
View Documents
N/A
2956
50 mm Staining Trough
16.05 €
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
12888735
View Documents
N/A
2960/1
50 mm Staining Jar
93.15 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Glass trough with lid
  • Holds microscope slides 76mm x 26mm
  • Two different styles
Specifications

Specifications

Closure Included
Flat
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.